Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC9C
Entrez Gene ID
483782
Gene Name
tetratricopeptide repeat domain 9C
Gene Aliases
-
Chromosome Location
Chr.18: 53928236 - 53941410 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005631611.3 XP_005631668.1 1 - 2 619 63
XM_022405782.1 XP_022261490.1 1 - 2 626 63
XM_022405783.1 XP_022261491.1 2 - 3 462 63
XM_022405784.1 XP_022261492.1 2 - 3 474 63
XM_540903.6 XP_540903.1 1 - 2 613 63

View Full Product Details