Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SOX10
Entrez Gene ID
481258
Gene Name
SRY-box transcription factor 10
Gene Aliases
-
Chromosome Location
Chr.10: 26680441 - 26690530 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_538379.6 XP_538379.3 1 - 2 192 130

View Full Product Details