Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RFK
Entrez Gene ID
484159
Gene Name
riboflavin kinase
Gene Aliases
-
Chromosome Location
Chr.1: 82062290 - 82071033 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005615844.3 XP_005615901.1 2 - 3 601 95

View Full Product Details