Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GADD45G
Entrez Gene ID
484198
Gene Name
growth arrest and DNA damage inducible gamma
Gene Aliases
-
Chromosome Location
Chr.1: 96745001 - 96746535 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_541314.6 XP_541314.3 2 - 3 268 57

View Full Product Details