Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAPBP
Entrez Gene ID
481740
Gene Name
TAP binding protein
Gene Aliases
TPN, TPSN
Chromosome Location
Chr.12: 2740276 - 2751808 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001048101.2 NP_001041566.1 5 - 6 1405 110
XM_022425733.1 XP_022281441.1 4 - 5 1136 110

View Full Product Details