Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBXL6
Entrez Gene ID
482094
Gene Name
F-box and leucine rich repeat protein 6
Gene Aliases
-
Chromosome Location
Chr.13: 37780991 - 37783976 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005628081.2 XP_005628138.1 3 - 4 646 92
XM_539215.5 XP_539215.2 3 - 4 644 92

View Full Product Details