Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLAGL2
Entrez Gene ID
485829
Gene Name
PLAG1 like zinc finger 2
Gene Aliases
-
Chromosome Location
Chr.24: 21594679 - 21608728 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_542953.6 XP_542953.2 1 - 2 128 110

View Full Product Details