Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VCPIP1
Entrez Gene ID
486980
Gene Name
valosin containing protein interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.29: 16204932 - 16235893 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_544109.6 XP_544109.2 1 - 2 3011 86

View Full Product Details