Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FDXACB1
Entrez Gene ID
489408
Gene Name
ferredoxin-fold anticodon binding domain containing 1
Gene Aliases
-
Chromosome Location
Chr.5: 21205520 - 21210629 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022418323.1 XP_022274031.1 - 361 63
XM_546526.6 XP_546526.2 - 361 63

View Full Product Details