Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FABP12
Entrez Gene ID
487017
Gene Name
fatty acid binding protein 12
Gene Aliases
-
Chromosome Location
Chr.29: 28693981 - 28701149 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_544146.5 XP_544146.2 1 - 2 135 64

View Full Product Details