Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RASL10A
Entrez Gene ID
486343
Gene Name
RAS like family 10 member A
Gene Aliases
-
Chromosome Location
Chr.26: 22620755 - 22623251 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_543469.5 XP_543469.2 2 - 3 592 70

View Full Product Details