Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLEC1A
Entrez Gene ID
486695
Gene Name
C-type lectin domain family 1 member A
Gene Aliases
-
Chromosome Location
Chr.27: 35896421 - 35920649 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014108678.2 XP_013964153.1 5 - 6 881 127

View Full Product Details