Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RERG
Entrez Gene ID
486665
Gene Name
RAS like estrogen regulated growth inhibitor
Gene Aliases
-
Chromosome Location
Chr.27: 31413464 - 31526838 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005637110.3 XP_005637167.1 3 - 4 281 92
XM_014108686.2 XP_013964161.1 4 - 5 447 92
XM_022411064.1 XP_022266772.1 4 - 5 496 92
XM_022411065.1 XP_022266773.1 3 - 4 163 92
XM_543792.6 XP_543792.3 4 - 5 466 92

View Full Product Details