Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TIGIT
Entrez Gene ID
487986
Gene Name
T cell immunoreceptor with Ig and ITIM domains
Gene Aliases
VSIG9, VSTM3
Chromosome Location
Chr.33: 18484106 - 18501517 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005639508.3 XP_005639565.2 2 - 3 2454 113

View Full Product Details