Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPM1F
Entrez Gene ID
486448
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1F
Gene Aliases
-
Chromosome Location
Chr.26: 31162810 - 31193586 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014108210.2 XP_013963685.1 5 - 6 836 73
XM_022410172.1 XP_022265880.1 5 - 6 808 73
XM_543574.6 XP_543574.3 5 - 6 1011 73

View Full Product Details