Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DDIT4L
Entrez Gene ID
487877
Gene Name
DNA damage inducible transcript 4 like
Gene Aliases
-
Chromosome Location
Chr.32: 22062630 - 22067364 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_544999.6 XP_544999.2 2 - 3 300 127

View Full Product Details