Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MUL1
Entrez Gene ID
487394
Gene Name
mitochondrial E3 ubiquitin protein ligase 1
Gene Aliases
-
Chromosome Location
Chr.2: 78394013 - 78402032 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_544520.5 XP_544520.2 1 - 2 231 113

View Full Product Details