Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASB18
Entrez Gene ID
486175
Gene Name
ankyrin repeat and SOCS box containing 18
Gene Aliases
-
Chromosome Location
Chr.25: 47026112 - 47087245 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022409847.1 XP_022265555.1 3 - 4 413 79

View Full Product Details