Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLCNKA
Entrez Gene ID
487422
Gene Name
chloride voltage-gated channel Ka
Gene Aliases
CLCNKB
Chromosome Location
Chr.2: 81612382 - 81627742 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_544547.5 XP_544547.2 11 - 12 1148 83

View Full Product Details