Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLDN7
Entrez Gene ID
489466
Gene Name
claudin 7
Gene Aliases
-
Chromosome Location
Chr.5: 32223985 - 32225795 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_546584.5 XP_546584.3 3 - 4 470 86

View Full Product Details