Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AQP5
Entrez Gene ID
486551
Gene Name
aquaporin 5
Gene Aliases
-
Chromosome Location
Chr.27: 4735313 - 4742975 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_543677.4 XP_543677.4 5 - 6 844 61

View Full Product Details