Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUDT1
Entrez Gene ID
489894
Gene Name
nudix hydrolase 1
Gene Aliases
-
Chromosome Location
Chr.6: 14874459 - 14881517 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_547012.5 XP_547012.4 - 275 76

View Full Product Details