Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNAPIN
Entrez Gene ID
490456
Gene Name
SNAP associated protein
Gene Aliases
SNAPAP
Chromosome Location
Chr.7: 43410554 - 43411929 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_547578.6 XP_547578.1 2 - 3 230 62

View Full Product Details