Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ERRFI1
Entrez Gene ID
489636
Gene Name
ERBB receptor feedback inhibitor 1
Gene Aliases
-
Chromosome Location
Chr.5: 61621964 - 61636152 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_546756.6 XP_546756.1 - 419 64

View Full Product Details