Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NTPCR
Entrez Gene ID
607237
Gene Name
nucleoside-triphosphatase, cancer-related
Gene Aliases
-
Chromosome Location
Chr.4: 6704571 - 6733820 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014112738.2 XP_013968213.2 - 627 83

View Full Product Details