Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCLRE1B
Entrez Gene ID
606778
Gene Name
DNA cross-link repair 1B
Gene Aliases
-
Chromosome Location
Chr.17: 51711267 - 51716806 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005630634.3 XP_005630691.1 3 - 4 800 67
XM_022404864.1 XP_022260572.1 3 - 4 495 67
XM_843181.5 XP_848274.1 3 - 4 800 67

View Full Product Details