Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM16
Entrez Gene ID
489518
Gene Name
tripartite motif containing 16
Gene Aliases
-
Chromosome Location
Chr.5: 39146505 - 39167857 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022418606.1 XP_022274314.1 3 - 4 678 73
XM_546637.5 XP_546637.2 3 - 4 899 73

View Full Product Details