Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARL4D
Entrez Gene ID
490950
Gene Name
ADP ribosylation factor like GTPase 4D
Gene Aliases
-
Chromosome Location
Chr.9: 19832608 - 19834593 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_548073.6 XP_548073.2 - 190 133

View Full Product Details