Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZG16B
Entrez Gene ID
490055
Gene Name
zymogen granule protein 16B
Gene Aliases
-
Chromosome Location
Chr.6: 38353189 - 38355163 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_547175.5 XP_547175.2 3 - 4 345 77

View Full Product Details