Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCL10
Entrez Gene ID
490183
Gene Name
BCL10 immune signaling adaptor
Gene Aliases
-
Chromosome Location
Chr.6: 62712937 - 62723852 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_547304.6 XP_547304.2 2 - 3 605 142

View Full Product Details