Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SERPINA4
Entrez Gene ID
490840
Gene Name
serpin family A member 4
Gene Aliases
-
Chromosome Location
Chr.8: 63552033 - 63560667 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005623769.2 XP_005623826.1 4 - 5 1185 69

View Full Product Details