Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPEB3
Entrez Gene ID
486801
Gene Name
cytoplasmic polyadenylation element binding protein 3
Gene Aliases
-
Chromosome Location
Chr.28: 6523894 - 6724010 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022411917.1 XP_022267625.1 10 - 11 2081 68
XM_022411918.1 XP_022267626.1 10 - 11 2082 68
XM_022411919.1 XP_022267627.1 9 - 10 2057 68
XM_022411921.1 XP_022267629.1 10 - 11 2011 68
XM_022411922.1 XP_022267630.1 9 - 10 1986 68
XM_022411923.1 XP_022267631.1 9 - 10 1936 68
XM_022411924.1 XP_022267632.1 9 - 10 1995 68
XM_022411925.1 XP_022267633.1 10 - 11 2022 68
XM_022411926.1 XP_022267634.1 9 - 10 1878 68
XM_022411927.1 XP_022267635.1 9 - 10 1869 68

View Full Product Details