Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMUB2
Entrez Gene ID
607070
Gene Name
transmembrane and ubiquitin like domain containing 2
Gene Aliases
-
Chromosome Location
Chr.9: 19204802 - 19209275 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005624403.3 XP_005624460.1 - 818 115
XM_005624405.3 XP_005624462.1 - 718 115
XM_014116520.2 XP_013971995.1 - 1266 115
XM_022422902.1 XP_022278610.1 - 1103 115
XM_022422903.1 XP_022278611.1 - 1359 115
XM_022422904.1 XP_022278612.1 - 1381 115
XM_022422905.1 XP_022278613.1 - 676 115
XM_843644.5 XP_848737.2 - 834 115

View Full Product Details