Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRSS22
Entrez Gene ID
490056
Gene Name
serine protease 22
Gene Aliases
-
Chromosome Location
Chr.6: 38371440 - 38375479 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022420233.1 XP_022275941.1 3 - 4 573 62

View Full Product Details