Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRPL55
Entrez Gene ID
607317
Gene Name
mitochondrial ribosomal protein L55
Gene Aliases
-
Chromosome Location
Chr.14: 826731 - 830107 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_843940.5 XP_849033.2 1 - 2 100 89

View Full Product Details