Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALKBH5
Entrez Gene ID
609383
Gene Name
alkB homolog 5, RNA demethylase
Gene Aliases
-
Chromosome Location
Chr.5: 41339292 - 41365641 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_846626.5 XP_851719.1 1 - 2 1863 95

View Full Product Details