Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NR2C2AP
Entrez Gene ID
609730
Gene Name
nuclear receptor 2C2 associated protein
Gene Aliases
-
Chromosome Location
Chr.20: 43976373 - 43978410 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_847056.5 XP_852149.1 3 - 4 531 103

View Full Product Details