Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC41
Entrez Gene ID
608535
Gene Name
leucine rich repeat containing 41
Gene Aliases
-
Chromosome Location
Chr.15: 14092739 - 14111034 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014119564.2 XP_013975039.1 3 - 4 734 84
XM_014119565.2 XP_013975040.1 3 - 4 269 84
XM_022427982.1 XP_022283690.1 3 - 4 645 84
XM_845204.5 XP_850297.1 3 - 4 644 84

View Full Product Details