Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MED28
Entrez Gene ID
488813
Gene Name
mediator complex subunit 28
Gene Aliases
-
Chromosome Location
Chr.3: 62887095 - 62895588 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005618542.3 XP_005618599.1 2 - 3 241 85
XM_022417147.1 XP_022272855.1 2 - 3 241 85
XM_845684.4 XP_850777.1 2 - 3 241 85

View Full Product Details