Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RASSF3
Entrez Gene ID
608312
Gene Name
Ras association domain family member 3
Gene Aliases
-
Chromosome Location
Chr.10: 7323272 - 7353378 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022424160.1 XP_022279868.1 - 158 89
XM_022424161.1 XP_022279869.1 - 158 89
XM_022424162.1 XP_022279870.1 - 232 89
XM_022424163.1 XP_022279871.1 - 218 89

View Full Product Details