Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYGB
Entrez Gene ID
609341
Gene Name
cytoglobin
Gene Aliases
-
Chromosome Location
Chr.9: 4191114 - 4201438 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077587.1 NP_001071055.1 - 157 77
XM_014116197.2 XP_013971672.1 - 316 77
GenBank mRNA DQ336161.1 - - 157 77

View Full Product Details