Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTHFSD
Entrez Gene ID
609306
Gene Name
methenyltetrahydrofolate synthetase domain containing
Gene Aliases
-
Chromosome Location
Chr.5: 66307852 - 66334196 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005620592.2 XP_005620649.1 - 451 120

View Full Product Details