Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRAP1
Entrez Gene ID
608254
Gene Name
proline rich acidic protein 1
Gene Aliases
-
Chromosome Location
Chr.28: 40927744 - 40931355 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_845222.5 XP_850315.1 4 - 5 367 89

View Full Product Details