Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DKKL1
Entrez Gene ID
610302
Gene Name
dickkopf like acrosomal protein 1
Gene Aliases
-
Chromosome Location
Chr.1: 107109787 - 107113932 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003638821.2 XP_003638869.1 3 - 4 497 80
XM_005616315.1 XP_005616372.1 3 - 4 479 80

View Full Product Details