Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KAZALD1
Entrez Gene ID
609313
Gene Name
Kazal type serine peptidase inhibitor domain 1
Gene Aliases
-
Chromosome Location
Chr.28: 13741085 - 13745426 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005637635.3 XP_005637692.2 3 - 4 1562 86

View Full Product Details