Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PVALB
Entrez Gene ID
481278
Gene Name
parvalbumin
Gene Aliases
-
Chromosome Location
Chr.10: 27655896 - 27674309 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003431505.3 XP_003431553.1 2 - 3 123 139
XM_022424476.1 XP_022280184.1 2 - 3 210 139
XM_022424477.1 XP_022280185.1 2 - 3 252 139
XM_022424478.1 XP_022280186.1 2 - 3 137 139
XM_538399.6 XP_538399.2 2 - 3 202 139

View Full Product Details