Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCAS4
Entrez Gene ID
611448
Gene Name
breast carcinoma amplified sequence 4
Gene Aliases
-
Chromosome Location
Chr.24: 37146001 - 37202277 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005635177.3 XP_005635234.1 3 - 4 356 108

View Full Product Details