Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC25A47
Entrez Gene ID
612516
Gene Name
solute carrier family 25 member 47
Gene Aliases
-
Chromosome Location
Chr.8: 68602525 - 68607364 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022422381.1 XP_022278089.1 2 - 3 159 104
XM_850243.5 XP_855336.2 3 - 4 282 104

View Full Product Details