Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBE2R2
Entrez Gene ID
611799
Gene Name
ubiquitin conjugating enzyme E2 R2
Gene Aliases
-
Chromosome Location
Chr.11: 50554173 - 50678284 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_849513.5 XP_854606.3 3 - 4 766 80

View Full Product Details