Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRABP2
Entrez Gene ID
612093
Gene Name
cellular retinoic acid binding protein 2
Gene Aliases
-
Chromosome Location
Chr.7: 41277580 - 41282158 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022421245.1 XP_022276953.1 2 - 3 435 70

View Full Product Details